missing translation for 'onlineSavingsMsg'
Learn More

Axygen 2.0mL Self-Standing Screw Cap Tubes

Features chemical resistant O-rings

Brand:  Axygen SCT-200-SS-G

Code : 68

Additional Details : Weight : 8.62000kg

 View more versions of this product

Product Code. 13158053

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.

  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification

Specifications

Specifications

RNase, DNase, pyrogen-free
Polypropylene
1.5mL
20000xG
Non-looped
O-ring
Green
Screw
Polypropylene
No
No
Conical
Videos