missing translation for 'onlineSavingsMsg'
Learn More

Axygen 2.0mL Self-Standing Screw Cap Tubes

Features chemical resistant O-rings

Brand:  Axygen SCT-200-SS-O

Code : A0

Additional Details : Weight : 8.20000kg

 View more versions of this product

Product Code. 11324195

  • £121.09 / Pack of 500


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.

  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification

Specifications

Specifications

RNase, DNase, pyrogen-free
Polypropylene
1.5mL
20000xG
Non-looped
O-ring
Orange
Screw
Polypropylene
No
No
Conical
Videos