missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 2.0mL Conical Screw Cap Tubes

Features O-ring resistant to many commonly-used laboratory chemicals

Brand:  Axygen SCT-200-GR

Code : 68

Additional Details : Weight : 8.20000kg

 View more versions of this product

Product Code. 12759568

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

RNase, DNase, pyrogen-free
Polypropylene
2mL
20000xG
Non-looped
O-ring
Gray
Screw
Polypropylene
No
No
Conical
Videos