missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 12-Strip PCR Tubes and Caps

Manufactured from 99.9% pure virgin polypropylene

Brand:  Axygen PCR-0212-C

Code : 68

Additional Details : Weight : 3.20000kg

 View more versions of this product

Product Code. 12706849

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Strips can be centrifuged at 4000 x g.

  • Ultra-thin and consistent wall thickness allows precise thermal transfer
  • Tubes of 0.2mL capacity

Specifications

Specifications

Thin Wall 12-Strip PCR Tube
Clear
0.2mL
No
4000xG
Thermal cyclers for thermal transfer applications
0.2mL
RNase, DNase-free
Polypropylene
80 Pack
Polypropylene
Strip
Conical
Videos