missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 1000μL Universal Pipette Tips: Wide Bore

Compatible with most popular brands of pipettes

Brand:  Axygen T-1005-WB-C-R-S

Code : A0

Additional Details : Weight : 13.83000kg

 View more versions of this product

Product Code. 12603716

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Orifice size of 0.065 in. (1.6mm) makes wide bore tips suitable for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Volume range: 100 to 1000μL
  • Available in bulk or racked formats

Specifications

Specifications

Rack
Rack
100 to 1000μL
Clear
Wide Bore
DNase, RNase, DNA, and PCR inhibitor Free
Universal Fit Pipettor
Genomic/Wide OrificeĆ¾Universal Fit
Sterile
Yes
PP (Polypropylene)
Universal Fit Pipettor
Videos