Learn More
Axygen™ Microvolume Extended Micropipette Tips
Axygen™ Microvolume Extended Micropipette Tips have an extended tip for easy access to narrow, deep wells.
Brand: Axygen™ TXL-10-L
Code : 68
Additional Details : Weight : 3.90000kg
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Microvolume
- Maxymum Recovery technology for ultra-low retention and high yield
Specifications
0.1 to 10 μL | |
Clear | |
DNase, Rnase Free | |
Maxymum Recovery™ surface | |
1,000 Tips/Pk., 20 Packs/Cs. | |
Polypropylene |
Non-sterile | |
USP 3127 | |
Allows recovery of costly samples with improved precision and accuracy | |
Low Retention, Extended Length, Microvolume | |
Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2 |