Learn More
Axygen™ 10µL Microvolume Filter Tips
Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, bevelled, Eppendorf-style, nonsterile, sterile, non-filtered, and other features for specialized applications.
Brand:  Axygen™ T-300-STK-S
Code : 68
Additional Details : Weight : 3.60000kg
Product Code. 12571298
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield
Specifications
Specifications
Tip
Sterile
Microvolume
Stack Rack
RNase, DNase-free
Comply with USP 3127 (Heavy Metal Contamination)
P2/P10 Gilson™ Pipetman™
0.5 to 10μL
Clear
Polypropylene
4800 Case
Allows recovery of costly samples with improved precision and accuracy
Videos