Learn More
Axygen™ 10µL Microvolume Filter Tips
Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, bevelled, Eppendorf-style, nonsterile, sterile, non-filtered, and other features for specialized applications.
Brand:  Axygen™ T-300-L-STK-S
Code : 68
Additional Details : Weight : 3.60000kg
Product Code. 12551298
  • £196.25 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield
Specifications
Specifications
Rack
Gilson™ Pipetman® P10þGilson™ Pipetman® P2
Low RetentionþMicrovolume
Sterile
PP (Polypropylene)
DNase, Rnase Free
Gilson™ Pipetman® P10, Gilson™ Pipetman® P2
4800 Case
Rack
0.5 to 10μL
Clear
USP 3127
Allows recovery of costly samples with improved precision and accuracy
Videos