Learn More
Axygen™ 10μL Microvolume Pipette Tips, Non-Filtered
Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes.
Brand: Axygen™ TXL-10-R
Code : 68
Additional Details : Weight : 8.16000kg
Description
- Polypropylene tips
- Microvolume
- Long length
- Maxymum Recovery technology for ultra-low retention
- Designed to fit a wide variety of single and multi-channel pipettes
- Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject
Specifications
0.1 to 10 μL | |
Non-sterile | |
USP 3127 | |
Rack | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Polypropylene |
Non-Filtered | |
Clear | |
Allows recovery of costly samples with improved precision and accuracy | |
Extended Length, Microvolume | |
Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2 |