Axygen™ 1.5mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Overview
Brand: Axygen
Manufacturer Part Number: SCT-150-SS-G
X500 1.5ML TUBE & CAP GREEN
UNSPSC: 41121702
Code: A0
Additional Details:
Additional Details: Weight: 8.62000kg
Product Code. 11390385
 N/A
N/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description and Specification
Specification
Capacity (Metric) 1.5mL
Material Polypropylene
Shape Conical
Closure Type Screw
Max. Force 20000xG
Sterile No
Format Non-looped
Graduated No
Includes O-ring
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Green
Closure Material Polypropylene