Learn More
Axygen™ 1.5mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Brand:  Axygen SCT-150-SS-B
Code : 68
Additional Details : Weight : 8.62000kg
 View more versions of this product
Product Code. 13167384
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
RNase, DNase, pyrogen-free
Polypropylene
1.5mL
20000xG
Non-looped
O-ring
Blue
Screw
Polypropylene
No
No
Conical
Videos