Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
Brand:  Axygen™ SCT-050-SS-A-S
Code : 68
Additional Details : Weight : 8.51000kg
 View more versions of this product
Product Code. 12779498
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile tubes
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Wide range of colored tubes for easy sample identification
Specifications
Specifications
20,000 x g
Assorted
0.5 mL
Non-looped
Bag
Screw Cap
RNase, DNase, pyrogen-free
Polypropylene
Polypropylene
O-ring
Sterile
Conical