Learn More
Axygen™ 0.6mL Conical Screw Cap Tubes: Presterilized
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents
Brand: Axygen™ SCT-45-C-S
Code : 68
Additional Details : Weight : 8.20000kg
Description
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
- Maximum G-force is 20,000 RCF.
- RNase-/DNase-free
- Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Specifications
20,000 x g | |
RNase, DNase, pyrogen-free | |
Polypropylene | |
Clear | |
Non-looped | |
500/Pk. | |
Sterile | |
Screw Cap |
Screw Cap | |
Clear | |
0.6 mL | |
Polypropylene | |
O-ring | |
500 Elangated Tubes and caps/Pk.; 8 Packs/Cs. | |
No | |
Conical |