Learn More
Axygen™ 0.5mL Conical Screw Cap Tubes: Nonsterile
Features chemical resistant O-ring
Brand:  Axygen SCT-050-R
Code : A0
Additional Details : Weight : 7.30000kg
 View more versions of this product
Product Code. 11320355
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Non-sterile
Red
Screw
Polypropylene
Non-looped
O-ring
Conical
RNase, DNase, pyrogen-free
Polypropylene
0.5mL
20000xG
No
500 Tubes and caps/Pk., 8 Pk./Cs.
Videos