missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 0.5mL Conical Screw Cap Tubes: Nonsterile
Features chemical resistant O-ring
Brand: Axygen SCT-050-C
Code : A0
Additional Details : Weight : 7.03000kg
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Available in range of colors for easy sample identification
Specifications
20,000 x g | |
Clear | |
0.5 mL | |
Non-looped | |
500 Tubes and caps/Pk., 8 Pk./Cs. | |
No | |
Conical |
RNase, DNase, pyrogen-free | |
Polypropylene | |
Polypropylene | |
O-ring | |
Non-sterile | |
Screw Cap |