Learn More
Axl Monoclonal Antibody (DS7HAXL), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£146.00 - £371.00
Specifications
Antigen | Axl |
---|---|
Clone | DS7HAXL |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963234
|
Affymetrix eBioscience
25-1087-42 |
100 Tests |
£371.00
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953234
|
Affymetrix eBioscience
25-1087-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This DS7HAXL monoclonal antibody recognizes human Axl, a member of the TAM family of tyrosine kinase receptors that also includes MerTK and Tyro3. This DS7HAXL antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. It has also been shown to stain formalin-fixed, paraffin-embedded tissues for IHC analysis. Applications Reported: This DS7HAXL antibody has been reported for use in flow cytometric analysis. Applications Tested: This DS7HAXL antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow Cytometry.Specifications
Axl | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P30530 | |
AXL | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
AXL |
DS7HAXL | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
558 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |