missing translation for 'onlineSavingsMsg'
Learn More

Axl Mouse anti-Human, APC, Clone: DS7HAXL, eBioscience™

Mouse Monoclonal Antibody

Brand:  Invitrogen 17-1087-41

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963204

  • £134.00 / Pack of 25
Estimated Shipment: 17-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This DS7HAXL monoclonal antibody recognizes human Axl, a member of the TAM family of tyrosine kinase receptors that also includes MerTK and Tyro3. This DS7HAXL antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. It has also been shown to stain formalin-fixed, paraffin-embedded tissues for IHC analysis. Applications Reported: This DS7HAXL antibody has been reported for use in flow cytometric analysis. Applications Tested: This DS7HAXL antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser

Axl is a member of the TAM receptor family, which also includes also MerTK and TYRO3. Axl contains protein kinase activity and is activated by dimerization and auto-phosphorylation upon ligand binding. In response to its ligands, Protein S and GAS6, Axl activates anti-inflammatory pathways (through SOCS-1 and SOCS-3), which limits TLR and cytokine signaling. This results in dampened inflammatory responses in macrophages and DCs. TAM receptors are also involved in phagocytosis of apoptotic cells. Mice lacking all three TAM receptors have several degenerative phenotypes linked to inefficient removal of apoptotic cells and membranes (e.g., in the retina and the male reproductive tract) and develop a severe autoimmune phenotype akin to systemic lupus erythematosus, including the production of broad spectrum auto-antibodies. In addition, Axl may function as a putative entry receptor for filoviruses. Axl is also used to identify a specific subpopulation of human blood dendritic cells, also referred to as Siglec 6+/Axl+ dendritic cells. Cellular expression of Axl can be upregulated by TLR ligands, such as LPS or poly I:C. Soluble Axl is generated by proteolytic cleavage of the membrane form. Increased plasma levels may be indicative of inflammation and cancer.
Specifications

Specifications

Axl
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P30530
ARK; AXL; JTK11; Tyro7; UFO
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DS7HAXL
APC
AXL
Liquid
AXL
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
558
Videos
SDS
Documents
Certificates
Special Offers

Special Offers