Learn More
Axl Monoclonal Antibody (DS7HAXL), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£341.00
Specifications
Antigen | Axl |
---|---|
Clone | DS7HAXL |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973204
|
Affymetrix eBioscience
17-1087-42 |
100 Tests |
£341.00
Pack of 100 |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This DS7HAXL monoclonal antibody recognizes human Axl, a member of the TAM family of tyrosine kinase receptors that also includes MerTK and Tyro3. This DS7HAXL antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. It has also been shown to stain formalin-fixed, paraffin-embedded tissues for IHC analysis. Applications Reported: This DS7HAXL antibody has been reported for use in flow cytometric analysis. Applications Tested: This DS7HAXL antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow Cytometry.Specifications
Axl | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P30530 | |
AXL | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
AXL |
DS7HAXL | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
558 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |