missing translation for 'onlineSavingsMsg'
Learn More

Axitinib, Tocris Bioscience™

Potent VEGFR-1, -2 and -3 inhibitor

Brand:  RD Systems (Bio-Techne) 4350/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13454604

  • £244.02 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent inhibitor of VEGFR-2, -3, and -1 (IC50 values are 0.2, 0.1-0.3, and 1.2 nM respectively). Exhibits minimal activity against a panel of ∽100 protein kinase. Inhibits angiogenesis and vascular permeability. Also a high affinity BCR-ABL1 (T315I) inhibitor (Ki = 149 pM for autophosphorylated ABL1 (T315I)). Blocks proliferation of Ba/F3 cells expressing BCR-ABL1 (T315I). Orally available.
Specifications

Chemical Identifiers

319460-85-0
386.473
axitinib, inlyta, axitinibum, unii-c9lvq0yuxg, e-n-methyl-2-3-2-pyridin-2-yl vinyl-1h-indazol-6-yl thio benzamide, c9lvq0yuxg, n-methyl-2-3-1e-2-pyridin-2-yl ethenyl-1h-indazol-6-yl sulfanyl benzamide, n-methyl-2-3-e-2-pyridin-2-yl ethenyl-1h-indazol-6-yl sulfanyl benzamide, e-n-methyl-2-3-2-pyridin-2-yl vinyl-1h-indazol-6-ylthio benzamide, dsstox_cid_28975
CHEBI:66910
CNC(=O)C1=CC=CC=C1SC2=CC3=C(C=C2)C(=NN3)C=CC4=CC=CC=N4
C22H18N4OS
RITAVMQDGBJQJZ-FMIVXFBMSA-N
6450551
N-methyl-2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]benzamide

Specifications

Axitinib
319460-85-0
axitinib, inlyta, axitinibum, unii-c9lvq0yuxg, e-n-methyl-2-3-2-pyridin-2-yl vinyl-1h-indazol-6-yl thio benzamide, c9lvq0yuxg, n-methyl-2-3-1e-2-pyridin-2-yl ethenyl-1h-indazol-6-yl sulfanyl benzamide, n-methyl-2-3-e-2-pyridin-2-yl ethenyl-1h-indazol-6-yl sulfanyl benzamide, e-n-methyl-2-3-2-pyridin-2-yl vinyl-1h-indazol-6-ylthio benzamide, dsstox_cid_28975
CNC(=O)C1=CC=CC=C1SC2=CC3=C(C=C2)C(=NN3)C=CC4=CC=CC=N4
386.473
CHEBI:66910
>99%
10mg
C22H18N4OS
RITAVMQDGBJQJZ-FMIVXFBMSA-N
N-methyl-2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]benzamide
6450551
386.47
Videos
Documents