Learn More
AVPI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558041
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASVVSEPPPW QAPIEARGRK QASANIFQDA ELLQIQGLFQ RSGDQLAEER AQIIWECAGD HRVAEALKR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 80%.
May be involved in MAP kinase activation, epithelial sodium channel (ENaC) down-regulation and cell cycling. Target protein is homologous to mouse arginine vasopressin-induced 1 gene.Specifications
AVPI1 | |
Polyclonal | |
Unconjugated | |
AVPI1 | |
arginine vasopressin induced 1; arginine vasopressin-induced 1; arginine vasopressin-induced protein 1; AVPI1; AVP-induced protein 1; NPD013; PP5395; RP11-548K23.7; vasopressin-induced transcript; VIP32; VIT32 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60370 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T686 | |
AVPI1 | |
Recombinant protein corresponding to Human AVPI1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only