Learn More
Molecular Probes™ Avidin, FITC conjugate
Avidin, FITC conjugate
Brand: Molecular Probes™ 434411
Code : 50
Additional Details : Weight : 0.02100kg
Description
Our avidin, fluorescein conjugate has excitation and emission maxima ~494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.Specifications
Store in refrigerator (2–8°C). | |
Wet Ice | |
FITC (Fluorescein) |
For Research Use Only. Not for use in diagnostic procedures.