Learn More
Novacyt - Microgen Bioproducts Avian Allergens Serological Double Gel Diffusion Assay
Microgen Bioproducts Avian Allergens - A serological double gel diffusion assay intended for the detection of precipitating antibodies to avian allergens
Brand:  Novacyt - Microgen Bioproducts FSK4
Additional Details : Weight : 0.50000kg
Product Code. 17343191
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Double diffusion assay for the detection of precipitating antibodies elicited to the inhalation of avian allergens.
Specifications
Specifications
6.5 mL
Double gel diffusion (Ouchterlony)
Test Kit
Liquid
Double Gel Diffusion Assay
5 to 10 mL of venous blood