Learn More
CORNING AXYGEN™ Automation Tips for Molecular Devices Flexstation
CORNING AXYGEN™ Automation Tips for Molecular Devices Flexstation are manufactured to stringent specifications for consistent quality.
Brand: CORNING AXYGEN™ RMT-200-BK-R
Code : NEW
Additional Details : Weight : 3.90000kg
Description
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
- Certified RNase-/DNase-free and nonpyrogenic.
Specifications
96-Tip Format | |
Non-Filtered | |
Clear | |
Automation |
200 μL | |
Non-sterile | |
RNase/DNase-free, nonpyrogenic |