missing translation for 'onlineSavingsMsg'
Learn More

Watson Marlow™ Autoclavable Thermoplastic Elastomer Transfer Tubing For 505Di/L Pumps

Use this tubing for a wide variety of applications. Watson Marlow™ Thermoplastic Tubing is resistant to a wide range of chemicals, offers long tubing life and is highly resistant to oxidizing agents such as ozone, peroxides and sodium hypochlorite.

Brand:  Watson Marlow™ 902.0032.024

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12446330

  • £252.00 / Each


Estimated Shipment
12-02-2018
Add to basket

Description

Description

Marprene is a long life tubing, highly resistant to attack by oxidising agents including ozone, peroxides and hypochlorite. It has high acid/alkali resistance and good resistance to animal and vegetable oils. Autoclavable, Marprene may be steam sterilized at 135°C or cold sterilized by ethylene oxide.
Specifications

Specifications

Yes
15m
Transfer Tubing
Yes
2.4mm
3.2mm
Yes
Thermoplastic Elastomer
Thermoplastic Elastomer
For 505Di/L Pump
1Each
Videos
Documentation