missing translation for 'onlineSavingsMsg'
Learn More

Watson Marlow™ Autoclavable Silicone Tubing For 505Di/L Pumps

Brand:  Watson Marlow™ 902.E016.K24

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12496330

  • £36.35 / Pack of 5


Estimated Shipment
15-03-2018
Add to basket

Description

Description

Marprene is a long life tubing, highly resistant to attack by oxidising agents including ozone, peroxides and hypochlorite. It has high acid/alkali resistance and good resistance to animal and vegetable oils. Autoclavable, Marprene may be steam sterilized at 135°C or cold sterilized by ethylene oxide.
Specifications

Specifications

Tubing
Yes
2.4mm
1.6mm
Yes
Silicone
For 505Di/L Pump
12Pack
Yes
Silicone
Videos
Documentation