Learn More
Duran™ Autoclavable Glass Screw-Thread Tubes With DIN 12257 and USP Standard
Brand:  Duran™ 248406202
Code : A0
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 13427467
  • £13.05 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
With GL14 DIN Thread and Standard Ground Cone
1Each
Screw-Thread Tube
Videos