missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Blender Bags

For autoclaving contaminated products safely

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 5.00000kg

Product Code. 11547762

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

77.9 x 40cm
78cm
40μm
21L
HDPE (High-density Polyethylene)
Clinical, dairy, environment, industries, microbiology
Biohazard symbol
Yes
Natural
40cm
Videos
Documentation