Learn More
Auranofin, Tocris Bioscience™
Thioredoxin reductase inhibitor; induces MPT
Brand:  RD Systems (Bio-Techne) 4600/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13444974
  • £76.95 / 10mg
Estimated Shipment
27-06-2019
Description
Description
Inhibitor of thioredoxin reductase (TrxR) (IC50 = 20 nM; Ki = 4 nM for the NADPH-reduced form of human cytosolic TrxR). Thought to induce the mitochondrial permeability transition via inhibition of mitochondrial TrxR. Also exhibits anti-inflammatory and immunosuppressive activities; inhibits 5-lipoxygenase at high concentrations and stimulates LTA hydrolase at low concentrations.
Specifications
Chemical Identifiers
34031-32-8
678.483
4-2-aminoethyl benzene sulfonyl fluoride hydrochloride
gold(1+);3,4,5-triacetyloxy-6-(acetyloxymethyl)oxane-2-thiolate;triethylphosphane
C20H34AuO9PS
AUJRCFUBUPVWSZ-UHFFFAOYSA-M
24199313
CCP(CC)CC.CC(=O)OCC1C(C(C(C(O1)[S-])OC(=O)C)OC(=O)C)OC(=O)C.[Au+]
Specifications
Auranofin
34031-32-8
4-2-aminoethyl benzene sulfonyl fluoride hydrochloride
CCP(CC)CC.CC(=O)OCC1C(C(C(C(O1)[S-])OC(=O)C)OC(=O)C)OC(=O)C.[Au+]
678.483
678.48
10mg
C20H34AuO9PS
AUJRCFUBUPVWSZ-UHFFFAOYSA-M
gold(1+);3,4,5-triacetyloxy-6-(acetyloxymethyl)oxane-2-thiolate;triethylphosphane
24199313
>98%
Videos