Learn More
ATP8B4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559122
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CSINGRIYGE VHDDLDQKTE ITQEKEPVDF SVKSQADREF QFFDHNLMES IKMGDPKVHE FLR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 73%.
ATP8B4 Polyclonal antibody specifically detects ATP8B4 in Human samples. It is validated for Immunohistochemistry.Specifications
ATP8B4 | |
Polyclonal | |
Unconjugated | |
ATP8B4 | |
ATP8B4; ATPase class I type 8B member 4; ATPase phospholipid transporting 8B4 (putative); ATPase, class I, type 8B, member 4; ATPIM; KIAA1939; P4-ATPase flippase complex alpha subunit ATP8B4; potential phospholipid-transporting ATPase IM; probable phospholipid-transporting ATPase IM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79895 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TF62 | |
ATP8B4 | |
Recombinant protein corresponding to Human ATP8B4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only