Learn More
ATP11A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557323
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SATGEIYLFC KGADSSIFPR VIEGKVDQIR ARVERNAVEG LRTLCVAYKR LIQEEYEGIC KLLQAAKVAL QDREKKLAEA YEQIEKDLTL LG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
ATP11A Polyclonal antibody specifically detects ATP11A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ATP11A | |
Polyclonal | |
Unconjugated | |
ATP11A | |
4930558F19Rik; ATP11A; ATPase 11A, class VI; ATPase 11A, p type; ATPase class VI type 11A; ATPase IS; ATPase phospholipid transporting 11A; ATPase, class 1, member h; ATPase, class VI, type 11A; Atpc1h; ATPIH; ATPIS; AU040868; Ih; KIAA1021; P4-ATPase flippase complex alpha subunit ATP11A; phospholipid-translocating ATPase; potential phospholipid-transporting ATPase IH; probable phospholipid-transporting ATPase IH; RP11-120K24.1; Ua20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23250 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P98196 | |
ATP11A | |
Recombinant protein corresponding to Human ATP11A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only