missing translation for 'onlineSavingsMsg'
Learn More

Atipamezole hydrochloride, Tocris Bioscience™

Selective α2 antagonist

Brand:  RD Systems (Bio-Techne) 2937/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13442084

  • £710.01 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective α2-adrenergic receptor antagonist (Ki values are 1.1, 1.0, 0.89, 1300 and 6500 nM for α2A, α2B, α2C, α1A and α1B receptors respectively). Brain penetrant.
Specifications

Chemical Identifiers

104075-48-1
248.754
atipamezole hydrochloride, atipamezole hcl, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-, monohydrochloride, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, pubchem21910, mpv1248 hydrochloride, mpv 1248 hydrochloride, mpv-1248 hydrochloride
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole;hydrochloride
C14H17ClN2
PCCVCJAQMHDWJY-UHFFFAOYSA-N
13649426
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3.Cl

Specifications

50mg
104075-48-1
atipamezole hydrochloride, atipamezole hcl, 1h-imidazole, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-, monohydrochloride, 4-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, 5-2-ethyl-2,3-dihydro-1h-inden-2-yl-1h-imidazole hydrochloride, pubchem21910, mpv1248 hydrochloride, mpv 1248 hydrochloride, mpv-1248 hydrochloride
CCC1(CC2=CC=CC=C2C1)C3=CN=CN3.Cl
248.754
248.75
Atipamezole hydrochloride
C14H17ClN2
PCCVCJAQMHDWJY-UHFFFAOYSA-N
5-(2-ethyl-1,3-dihydroinden-2-yl)-1H-imidazole;hydrochloride
13649426
>99%
Videos
Documents