missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG5 (Autophagy Marker) Mouse, Clone: ATG5/2101, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 9474-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
ATG5 (Autophagy Marker) | |
Cytoplasmic | |
ATG5/2101 | |
Unconjugated | |
Mouse | |
50-32kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
9474 | |
IgG1 κ |
ELISA, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human ATG5 protein (apprx. aa 1-119) | |
THP-1, PANC-1, Raji or HeLa cells. Brain, Ovary, Endometrium, Colon or Duodenum | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |