missing translation for 'onlineSavingsMsg'
Learn More

ATC 0175 hydrochloride, Tocris Bioscience™

MCH1 antagonist; also 5-HT2B antagonist and 5-HT1A partial agonist

Brand:  Tocris (Bio-Techne) 334610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13442854

  • £210.21 / 10mg

Estimated Shipment date
from Supplier 19-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective and orally active melanin-concentrating hormone receptor 1 (MCH1) antagonist (IC50 values are 13.5 and > 10000 nM for MCH1 and MCH2 receptors respectively). Exhibits anxiolytic and antidepressant activity in rodents. Also displays affinity for 5-HT2B and 5-HT1A receptors (IC50 values are 9.66 and 16.9 nM respectively).
Specifications

Chemical Identifiers

510733-97-8
461.942
atc 0175 hydrochloride, n-cis-4-4-dimethylamino quinazolin-2-yl amino cyclohexyl-3,4-difluorobenzamide hcl, n-cis-4-4-dimethylamino-2-quinazolinyl amino cyclohexyl-3,4-difluorobenzamide hydrochloride, benzamide, n-cis-4-4-dimethylamino-2-quinazolinyl amino cyclohexyl-3,4-difluoro-, hydrochloride 1:1, benzamide, n-cis-4-4-dimethylamino-2-quinazolinyl amino cyclohexyl-3,4-difluoro-, monohydrochloride, atc hplc , solid, 3,4-difluoro-n-1s,4s-4-4-dimethylamino quinazolin-2-yl amino cyclohexyl benzamide hydrochloride
N-[4-[[4-(dimethylamino)quinazolin-2-yl]amino]cyclohexyl]-3,4-difluorobenzamide;hydrochloride
C23H26ClF2N5O
HUUPKFUYSQNNLO-UHFFFAOYSA-N
9934032
CN(C)C1=NC(=NC2=CC=CC=C21)NC3CCC(CC3)NC(=O)C4=CC(=C(C=C4)F)F.Cl

Specifications

10mg
510733-97-8
atc 0175 hydrochloride, n-cis-4-4-dimethylamino quinazolin-2-yl amino cyclohexyl-3,4-difluorobenzamide hcl, n-cis-4-4-dimethylamino-2-quinazolinyl amino cyclohexyl-3,4-difluorobenzamide hydrochloride, benzamide, n-cis-4-4-dimethylamino-2-quinazolinyl amino cyclohexyl-3,4-difluoro-, hydrochloride 1:1, benzamide, n-cis-4-4-dimethylamino-2-quinazolinyl amino cyclohexyl-3,4-difluoro-, monohydrochloride, atc hplc , solid, 3,4-difluoro-n-1s,4s-4-4-dimethylamino quinazolin-2-yl amino cyclohexyl benzamide hydrochloride
CN(C)C1=NC(=NC2=CC=CC=C21)NC3CCC(CC3)NC(=O)C4=CC(=C(C=C4)F)F.Cl
461.942
461.94
ATC 0175 hydrochloride
C23H26ClF2N5O
HUUPKFUYSQNNLO-UHFFFAOYSA-N
N-[4-[[4-(dimethylamino)quinazolin-2-yl]amino]cyclohexyl]-3,4-difluorobenzamide;hydrochloride
9934032
>98%
Videos
Documents