missing translation for 'onlineSavingsMsg'
Learn More

AT 1015, Tocris Bioscience™

Long-acting 5-HT2A antagonist

Brand:  RD Systems (Bio-Techne) 4468/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13434804

  • £546.84 / 50mg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Long-acting 5-HT2A receptor antagonist; inhibits vasoconstriction and blocks platelet aggregation. Prolongs clotting time in a rat model of thrombus formation; ameliorates laurate-induced peripheral vascular lesions in rodents.
Specifications

Chemical Identifiers

190508-50-0
492.06
at-1015 anhydrous, unii-oo0x01e9mj, oo0x01e9mj, 4-piperidinecarboxamide, n-2-4-5h-dibenzo a,d cyclohepten-5-ylidene-1-piperidinyl ethyl-1-formyl-, hydrochloride 1:1, n-2-4-5h-dibenzo a,d cyclohepten-5-ylidene-1-piperidinyl ethyl-1-formyl-4-piperidinecarboxamide hydrochloride
N-[2-[4-(dibenzo[1,2-a
C29H34ClN3O2
YZHGXYNLMHGNJZ-UHFFFAOYSA-N
9805663
C1CN(CCC1C(=O)NCCN2CCC(=C3C4=CC=CC=C4C=CC5=CC=CC=C53)CC2)C=O.Cl

Specifications

50mg
190508-50-0
at-1015 anhydrous, unii-oo0x01e9mj, oo0x01e9mj, 4-piperidinecarboxamide, n-2-4-5h-dibenzo a,d cyclohepten-5-ylidene-1-piperidinyl ethyl-1-formyl-, hydrochloride 1:1, n-2-4-5h-dibenzo a,d cyclohepten-5-ylidene-1-piperidinyl ethyl-1-formyl-4-piperidinecarboxamide hydrochloride
C1CN(CCC1C(=O)NCCN2CCC(=C3C4=CC=CC=C4C=CC5=CC=CC=C53)CC2)C=O.Cl
492.06
492.05
AT 1015
C29H34ClN3O2
YZHGXYNLMHGNJZ-UHFFFAOYSA-N
N-[2-[4-(dibenzo[1,2-a
9805663
>97%
Videos
Documents