missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASXL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565126
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PKSMPVSEASL IRIVPVVSQS ECKEEALQMS SPGRKEECES QGEVQPNFST SSEPLLSSAL NTHELSSILP IKCPKDE Highest antigen sequence identity to the following orthologs - mouse 62%, rat 54%.
ASXL2 Polyclonal antibody specifically detects ASXL2 in Human samples. It is validated for Immunocytochemistry.Specifications
ASXL2 | |
Polyclonal | |
Unconjugated | |
ASXL2 | |
4930556B16Rik; additional sex combs like 2; additional sex combs like 2 (Drosophila); additional sex combs like 2, transcriptional regulator; additional sex combs like transcriptional regulator 2; additional sex combs-like protein 2; ASXH2; ASXL transcriptional regulator 2; ASXL2; Kiaa1685; mKIAA1685; polycomb group protein ASXH2; polycomb group protein ASXH2 homolog; putative Polycomb group protein ASXL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55252 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q76L83 | |
ASXL2 | |
Recombinant Human ASXL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only