missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain AstiPure™ TP1 Series PFA Union Tee Fittings

Brand:  Saint-Gobain TP10406

Code : A0

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 11999934

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Union Tee Fittings
PFA
For 4 x 6mm dia. tubing
38 mm
4 mm
62 mm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers