Learn More
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Brand: Thermo Scientific™ C4013-71A
Code : 17
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
- Open top caps can be used with any 8mm septa
- Phenolic caps are suitable for autoclaving and low temperature applications
- Polypropylene caps are chemically inert and suitable for most chromatography applications
- Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
- PTFE/red rubber septa are economical for many GC applications
- PTFE/silicone septa are most popular for HPLC and IC applications
- Pre-slit septa are easier to pierce with thin-gauge needles
Specifications
Phenolic, Black; Ivory PTFE/Red Rubber septum | |
PTFE, Rubber, Phenolic | |
Open Top Cap | |
8-425 Standard Opening Vials |
Black | |
Ivory PTFE/Red Rubber | |
8-425 |