missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Aspirator Bottles without Stoppers

Brand:  Duran™ 247017307

Code : NEW

Additional Details : Weight : 1.81000kg

 View more versions of this product

Product Code. 10312751

  • £93.02 / Each


Estimated Shipment
19-02-2018
Add to basket

Specifications

Specifications

5000mL
320mm
NS 29/32
Aspirator Bottle
Videos