Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, complete with Glass Stopcock and Glass Stopper
Aspirator bottle, complete with glass stopcock and glass stopper.
Brand: DWK Life Sciences 247025401
Code : NEW
Additional Details : Weight : 6.00000kg
Description
Aspirator bottles, complete with glass stopcock. Complying with ISO 4796, these aspirators have a ground joint neck and glass stopper
- Supplied ready to use
- DURAN™ glass stopcock
- DURAN™ glass stopper
- Large range of sizes available
Specifications
107 mm | |
Borosilicate Glass (3.3) | |
Standard Taper Joint Bottom 19/26 | |
Autoclavable |
1000 mL | |
Storage and dispensing of liquids and reagents | |
10/Pk. |