Learn More
Tocris Bioscience™ Asenapine maleate
Non-selective 5-HT, dopamine, adrenalin and histamine receptor antagonist; antipsychotic agent
Brand:  Tocris Bioscience™ 373750
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433524
  • £673.26 / 50mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Novel psychopharmacologic agent. Displays antagonist activity at 5-HT, dopamine, noradrenalin and histamine receptor subtypes (pKi values are 8.60, 8.40, 10.15, 9.75, 10.46, 8.84, 9.60, 9.94, 8.85, 8.90, 8.84, 9.38, 8.95, 8.93, 8.9, 9.49, 8.91, 9.00 and 8.21 for 5-HT1A, 5-HT1B, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT5A, 5-HT6, 5-HT7, D1, D2L, D2S, D3, D4, α1A, α2A, α2B, α2C, H1 and H2 receptors respectively). Displays no appreciable affinity for muscarinic receptors. Exhibits potent activity in animal models predictive of antipsychotic efficacy.
Specifications
Specifications
Asenapine maleate
>98
50mg
Asenapine maleate
401.84