missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid, Fine Powder, 200 to 325 Mesh, U.S.P. - F.C.C., J.T.Baker™

Brand:  JT Baker 0937-08

Additional Details : CAS Number : 50-81-7 Weight : 2.90000kg

Packaging Poly Bottle
Quantity 1kg

 View more versions of this product

Product Code. 15587874

  • £800.03 / 1kg

Estimated Shipment
15-12-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

50-81-7
176.124
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
C(C(C1C(=C(C(=O)O1)O)O)O)O
C6H8O6
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one

Specifications

Ascorbic Acid
Poly Bottle
1kg
C6H8O6
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.12
FCC/USP
200 to 325 Mesh
1L = 1.65kg
50-81-7
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
C(C(C1C(=C(C(=O)O1)O)O)O)O
176.124
CHEBI:29073
Fine Powder
Videos
Documents