missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, Fine Powder, 200 to 325 Mesh, Multi-Compendial, U.S.P., J.T. Baker™
1610.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Mesh Size | 200 to 325 Mesh |
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 1 kg |
Specific Gravity | 1L = 1.65kg |
Formula Weight | 176.12 |
Grade | Multi-Compendial/USP |
Physical Form | Fine Powder |
Chemical Name or Material | Ascorbic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'