missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, Fine Granular, 20 to 80 Mesh, U.S.P. - F.C.C., J.T. Baker™
1201.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
125 g
500 g
1 kg
Packaging:
Poly Bottle
Specifications
Mesh Size | 20 to 80 Mesh |
Packaging | Poly Bottle |
Quantity | 1 kg |
Specific Gravity | 1L = 1.65kg |
Formula Weight | 176.12 |
Grade | FCC/USP |
Physical Form | Fine Granular |
Chemical Name or Material | Ascorbic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'