Learn More
ASCC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556796
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QLNNMDEVCY ENVLKQVKAG HQVMVFVHAR NATVRTAMSL IERAKNCGHI PFFFPTQGHD YVLAEKQVQR SRNKQVRELF PDGFSIHHAG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
ASCC3 Polyclonal antibody specifically detects ASCC3 in Human samples. It is validated for Immunohistochemistry.Specifications
ASCC3 | |
Polyclonal | |
Unconjugated | |
ASCC3 | |
Activating signal cointegrator 1 complex subunit 3; ASC-1 complex subunit P200; ASC1p200; ASCC3; B630009I04Rik; BC023451; D430001L07Rik; D630041L21; dJ121G13.4; dJ467N11.1; Helic1; Helicase, ATP binding 1; RNA helicase family; RNAH; Trip4 complex subunit p200 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10973 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3C0 | |
ASCC3 | |
Recombinant protein corresponding to Human ASCC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only