Learn More
Tocris Bioscience™ AS 605240
Potent and selective PI 3-kinase γ (PI 3-Kγ) inhibitor
Brand:  Tocris Bioscience™ 357850
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433234
  • £646.80 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Potent and selective inhibitor of PI 3-kinase γ (PI 3-Kγ) (IC50 = 8 nM). Displays 30-fold selectivity over PI 3-Kδ and PI 3-Kβ and 7.5-fold selectivity over PI 3-Kα. Suppresses the progression of joint inflammation and damage in both lymphocyte-independent and lymphocyte-dependent mouse models of rheumatoid arthritis. Orally active and ATP-competitive.
Specifications
Specifications
AS 605240
>98
50mg
AS 605240
257.27