missing translation for 'onlineSavingsMsg'
Learn More

AS 19, Tocris Bioscience™

Potent 5-HT7 agonist

Brand:  Tocris (Bio-Techne) 1968/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13440324

  • £207.20 / 10mg

Estimated Shipment
09-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent 5-HT7 receptor agonist (IC50 = 0.83 nM). Enhances memory consolidation and reverses scopolamine- or dizocilpine-induced amnesia in vivo.
Specifications

Chemical Identifiers

1000578-26-6
283.419
as-19 drug, d0x9ys, 2s-+-5-1,3,5-trimethylpyrazol-4-yl-2-dimethylamino tetralin, 2s-n,n-dimethyl-5-1,3,5-trimethylpyrazol-4-yl-1,2,3,4-tetrahydronaphthalen-2-amine
(2S)-N,N-dimethyl-5-(1,3,5-trimethylpyrazol-4-yl)-1,2,3,4-tetrahydronaphthalen-2-amine
C18H25N3
BTTOYOKCLDAHHO-HNNXBMFYSA-N
23642275
CC1=C(C(=NN1C)C)C2=C3CCC(CC3=CC=C2)N(C)C

Specifications

AS 19
1000578-26-6
as-19 drug, d0x9ys, 2s-+-5-1,3,5-trimethylpyrazol-4-yl-2-dimethylamino tetralin, 2s-n,n-dimethyl-5-1,3,5-trimethylpyrazol-4-yl-1,2,3,4-tetrahydronaphthalen-2-amine
CC1=C(C(=NN1C)C)C2=C3CCC(CC3=CC=C2)N(C)C
283.419
283.41
10mg
C18H25N3
BTTOYOKCLDAHHO-HNNXBMFYSA-N
(2S)-N,N-dimethyl-5-(1,3,5-trimethylpyrazol-4-yl)-1,2,3,4-tetrahydronaphthalen-2-amine
23642275
>98%
Videos
Documents