missing translation for 'onlineSavingsMsg'
Learn More

Arvanil, Tocris Bioscience™

Potent CB1 and TRPV1 agonist. Also anandamide transport inhibitor

Brand:  Tocris (Bio-Techne) 13545

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13439213

  • £64.68 / 5mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cannabinoid CB1 and vanilloid TRPV1 (VR1) agonist (Ki values are 0.5 and 0.3 μM respectively). Also inhibits the anandamide transporter (IC50 = 3.6 μM). Analgesic, vasodilatory and anti-inflammatory in vivo.
Specifications

Chemical Identifiers

128007-31-8
439.64
arvanil, n-vanillylarachidonamide, nava, n-arachidonoyl vanillylamine, n-4-hydroxy-3-methoxyphenyl methyl-5,8,11,14-eicosatetraenamide all-z, 5,8,11,14-eicosatetraenamide, n-4-hydroxy-3-methoxyphenyl methyl-, all-z, n-5z,8z,11z,15z-eicosatetraenoyl-vanillylamine, n-4-hydroxy-3-methoxyphenyl methyl-5z,8z,11z,14z-eicosatetraenamide, 5z,8z,11z,14z-n-4-hydroxy-3-methoxyphenyl methyl icosa-5,8,11,14-tetraenamide, d0d7qh
(5Z,8Z,11Z,14Z)-N-[(4-hydroxy-3-methoxyphenyl)methyl]icosa-5,8,11,14-tetraenamide
C28H41NO3
QVLMCRFQGHWOPM-ZKWNWVNESA-N
6449767
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC1=CC(=C(C=C1)O)OC

Specifications

5mg
128007-31-8
arvanil, n-vanillylarachidonamide, nava, n-arachidonoyl vanillylamine, n-4-hydroxy-3-methoxyphenyl methyl-5,8,11,14-eicosatetraenamide all-z, 5,8,11,14-eicosatetraenamide, n-4-hydroxy-3-methoxyphenyl methyl-, all-z, n-5z,8z,11z,15z-eicosatetraenoyl-vanillylamine, n-4-hydroxy-3-methoxyphenyl methyl-5z,8z,11z,14z-eicosatetraenamide, 5z,8z,11z,14z-n-4-hydroxy-3-methoxyphenyl methyl icosa-5,8,11,14-tetraenamide, d0d7qh
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC1=CC(=C(C=C1)O)OC
439.64
439.64
Arvanil
C28H41NO3
QVLMCRFQGHWOPM-ZKWNWVNESA-N
(5Z,8Z,11Z,14Z)-N-[(4-hydroxy-3-methoxyphenyl)methyl]icosa-5,8,11,14-tetraenamide
6449767
>98%
Videos
Documents