missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Artemin Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | Artemin |
---|---|
Clone | 314908 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Artemin Monoclonal specifically detects Artemin in Human samples. It is validated for Western Blot.Specifications
Artemin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q5T4W7.1 | |
9048 | |
E. coli-derived recombinant human Artemin Ala108-Gly220 Accession # Q5T4W7.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Artemin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BIG-H3, rhCripto-1, recombinant mouse (rm) Cripto-1, recombinant Drosophila DPP, rhDPP-6, rhGDNF, rhLAP, rhLatent TGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 3, rhLefty, rmLefty-1, rhMIS, rhNeurturin, rmNODAL, rhPersephin, recombinant amphibian TGF-beta 5, rhTGF-alpha or rhTGF-beta 2 is observed. |
314908 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin | |
ARTN | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only