Learn More
Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Hinged Rack.
Brand: Thermo Scientific™ 2921-HR
Code : A0
Additional Details : Weight : 1.79100kg
Description
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks.
Features:
- ART barrier tips provide 100% security against aerosols and liquid contamination
- Racks offer quick and easy access to pipette tips, saving valuable time
- Front latch closes and locks, ensuring safe and secure storage throughout
Certifications:
- Nonsterile products certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
- Sterile products certified to be free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
Recommended for:
- Liquid Handling
- Molecular Biology
- PCR
- Biotechnology
Specifications
Hinged Racks | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
ART |
ART Essentials Kit | |
Hinged Rack | |
Sterile & PCR Ready |