Learn More
Thermo Scientific™ ART Filtered Pipette Tips in Racks in Lift-off Lids
Pipette samples with the accuracy and precision you expect from your pipette, every time with SoftFit-L™ Filtered Pipette Tips in Racks in Lift-off Lids. For use with Rainin™ LTS pipettes.
Brand: Thermo Scientific™ 2275
Code : 03
Additional Details : Weight : 0.10000kg
Description
The SoftFit-L System is compatible with Rainin LTS pipettes made by Rainin Instrument LCC.
- Available in low retention
- Sterile tips are certified free from RNase, DNase and pyrogen
- Manufactured according to the industry's highest quality standards
Specifications
500 μL | |
Sterile | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Rainin LTS LiteTouch | |
Sterile & PCR Ready |
Filtered | |
Clear | |
Rack | |
ART |